siRNA Details
| Gene Symbol: TTTY22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001539.1 | 4, 5 | 387 | |||
| GenBank mRNA | AF527834.1 | - | 4, 5 | 387 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTTY22| Gene Symbol: TTTY22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001539.1 | 4, 5 | 387 | |||
| GenBank mRNA | AF527834.1 | - | 4, 5 | 387 | ||