siRNA Details
| Gene Symbol: FAM215A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026770.1 | 1 | 437 | |||
| GenBank mRNA | AF143236.1 | - | 1 | 449 | ||
| BC069097.1 | - | 1 | 437 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM215A| Gene Symbol: FAM215A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026770.1 | 1 | 437 | |||
| GenBank mRNA | AF143236.1 | - | 1 | 449 | ||
| BC069097.1 | - | 1 | 437 | |||