siRNA Details
| Gene Symbol: FRG2DP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026980.2 | 3 | 568 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FRG2DP| Gene Symbol: FRG2DP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026980.2 | 3 | 568 | |||