siRNA Details
| Gene Symbol: TRPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002720.2 | 1 | 105 | |||
| GenBank mRNA | AA423920.1 | - | 1 | 104 | ||
| X89067.1 | - | 1 | 88 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRPC2| Gene Symbol: TRPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002720.2 | 1 | 105 | |||
| GenBank mRNA | AA423920.1 | - | 1 | 104 | ||
| X89067.1 | - | 1 | 88 | |||