siRNA Details
Gene Symbol: PPY2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002181.1 | 1 | 338 | |||
GenBank mRNA | AF085945.1 | - | 1 | 233 | ||
AF222903.1 | - | 1 | 338 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PPY2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002181.1 | 1 | 338 | |||
GenBank mRNA | AF085945.1 | - | 1 | 233 | ||
AF222903.1 | - | 1 | 338 |