siRNA Details
| Gene Symbol: LOC646214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027053.2 | 1 | 6515 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC646214| Gene Symbol: LOC646214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027053.2 | 1 | 6515 | |||