siRNA Details
| Gene Symbol: RBMY3AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001573.2 | 1 | 22 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | U94386.3 | - | 1 | 22 | Chr. Y - 9610721 - 9615153 [-] on Build GRCh38 | |

RBMY3AP
Genomic Map