siRNA Details
| Gene Symbol: PAX8-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015377.2 | 6 | 2422 | |||
| GenBank mRNA | AK001856.1 | - | 3 | 1570 | ||
| AK056355.1 | - | 6 | 2060 | |||
| AL390179.1 | - | 3 | 1580 | |||
| BC033562.1 | - | 5 | 2305 | |||
| BC036699.2 | - | 6 | 2422 | |||
| BC042373.1 | - | 2 | 442 | |||
| BX537688.1 | - | 5 | 1963 | |||

PAX8-AS1
Genomic Map