siRNA Details
| Gene Symbol: PACRG-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028390.1 | 3 | 923 | |||
| GenBank mRNA | AK094729.1 | - | 3 | 911 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PACRG-AS1| Gene Symbol: PACRG-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028390.1 | 3 | 923 | |||
| GenBank mRNA | AK094729.1 | - | 3 | 911 | ||