siRNA Details
| Gene Symbol: BCAR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024049.1 | 4 | 1253 | |||
| NR_024050.1 | 3 | 611 | ||||
| NR_131216.1 | 2 | 541 | ||||
| NR_131217.1 | 3 | 826 | ||||
| NR_131222.1 | 3 | 659 | ||||
| GenBank mRNA | BX107175.1 | - | 3 | 582 | ||

BCAR4
Genomic Map