siRNA Details
| Gene Symbol: LOC154761 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015421.1 | 3 | 1027 | |||
| GenBank mRNA | AK124492.1 | - | 1 | 1790 | ||
| BC041464.1 | - | 2 | 390 | |||
| Gene Symbol: LOC101928422 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017012897.1 | XP_016868386.1 | 5 | 1460 | ||

LOC154761
Genomic Map