siRNA Details
| Gene Symbol: RPS26P11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002309.1 | 1 | 91 | |||
| GenBank mRNA | AW972305.1 | - | 1 | 201 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPS26P11| Gene Symbol: RPS26P11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002309.1 | 1 | 91 | |||
| GenBank mRNA | AW972305.1 | - | 1 | 201 | ||