siRNA Details
| Gene Symbol: PI4KAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003563.1 | 12 | 1762 | |||
| GenBank mRNA | AK002141.1 | - | 1 | 908 | ||
| AK024034.1 | - | 1 | 982 | |||
| BC131538.1 | - | 12 | 1762 | |||
| Gene Symbol: PI4KA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK095811.1 | - | 11 | 1484 | ||
| AK124692.1 | - | 8 | 2409 | |||

PI4KAP1
Genomic Map