siRNA Details
| Gene Symbol: CARMN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126481.1 | - | 5 | 2060 | ||
| AL137734.1 | - | 1 | 2399 | |||
| CR933654.1 | - | 2 | 5024 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CARMN| Gene Symbol: CARMN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126481.1 | - | 5 | 2060 | ||
| AL137734.1 | - | 1 | 2399 | |||
| CR933654.1 | - | 2 | 5024 | |||