siRNA Details
| Gene Symbol: CARMN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093957.1 | - | 1 | 858 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CARMN| Gene Symbol: CARMN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093957.1 | - | 1 | 858 | ||