siRNA Details
| Gene Symbol: LINC00174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026873.1 | 5 | 4161 | |||
| GenBank mRNA | AK091213.1 | - | 1 | 2819 | ||
| AK092625.1 | - | 1 | 1938 | |||
| AK124597.1 | - | 1 | 3282 | |||
| BQ645787.1 | - | 1 | 174 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00174| Gene Symbol: LINC00174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026873.1 | 5 | 4161 | |||
| GenBank mRNA | AK091213.1 | - | 1 | 2819 | ||
| AK092625.1 | - | 1 | 1938 | |||
| AK124597.1 | - | 1 | 3282 | |||
| BQ645787.1 | - | 1 | 174 | |||