siRNA Details
| Gene Symbol: URAHP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027335.2 | 3 | 334 | |||
| NR_027336.2 | 3 | 287 | ||||
| GenBank mRNA | AK304171.1 | - | 3 | 277 | ||
| BC044232.1 | - | 3 | 334 | |||
| BC089450.1 | - | 3 | 270 | |||
| DC393803.1 | - | 3 | 288 | |||
| Gene Symbol: GAS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC036781.1 | - | 1 | 357 | ||

URAHP
Genomic Map