siRNA Details
| Gene Symbol: GAS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC036781.1 | - | 1 | 266 | ||
| Gene Symbol: URAHP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027335.2 | 3 | 243 | |||
| NR_027336.2 | 3 | 196 | ||||
| GenBank mRNA | AK304171.1 | - | 3 | 186 | ||
| BC044232.1 | - | 3 | 243 | |||
| BC089450.1 | - | 3 | 179 | |||
| DC393803.1 | - | 3 | 197 | |||

GAS8
Genomic Map