siRNA Details
| Gene Symbol: URAHP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027335.2 | 4 | 536 | |||
| NR_027336.2 | 4 | 489 | ||||
| GenBank mRNA | AK304171.1 | - | 4 | 479 | ||
| BC044232.1 | - | 4 | 536 | |||
| BC089450.1 | - | 4 | 472 | |||
| DC393803.1 | - | 4 | 490 | |||
| Gene Symbol: GAS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC036781.1 | - | 1 | 1071 | ||

URAHP
Genomic Map