siRNA Details
| Gene Symbol: TPTE2P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026730.1 | 5 | 3517 | |||
| GenBank mRNA | AK126328.1 | - | 2 | 3420 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TPTE2P1| Gene Symbol: TPTE2P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026730.1 | 5 | 3517 | |||
| GenBank mRNA | AK126328.1 | - | 2 | 3420 | ||