siRNA Details
| Gene Symbol: WEE2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015392.1 | 4, 5 | 543 | |||
| GenBank mRNA | AK098171.1 | - | 4, 5 | 543 | ||
| BC101359.2 | - | 4, 5 | 797 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WEE2-AS1| Gene Symbol: WEE2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015392.1 | 4, 5 | 543 | |||
| GenBank mRNA | AK098171.1 | - | 4, 5 | 543 | ||
| BC101359.2 | - | 4, 5 | 797 | |||