siRNA Details
| Gene Symbol: WEE2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015392.1 | 7 | 1062 | |||
| GenBank mRNA | AK098171.1 | - | 7 | 1062 | ||
| BC101358.2 | - | 4 | 506 | |||
| BC101359.2 | - | 7 | 1316 | |||
| BC101360.2 | - | 3 | 352 | |||
| BC131564.1 | - | 5 | 692 | |||

ENSG00000228775
Genomic Map