siRNA Details
| Gene Symbol: WEE2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015392.1 | 7 | 1359 | |||
| GenBank mRNA | AK098171.1 | - | 7 | 1359 | ||
| BC101358.2 | - | 4 | 803 | |||
| BC101359.2 | - | 7 | 1613 | |||
| BC101360.2 | - | 3 | 649 | |||
| BC131564.1 | - | 5 | 989 | |||

WEE2-AS1
Genomic Map