siRNA Details
| Gene Symbol: LINC00909 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024484.1 | 1 | 931 | |||
| GenBank mRNA | AK001853.1 | - | 1 | 678 | ||
| BC036588.2 | - | 1 | 922 | |||
| BC132930.1 | - | 1 | 707 | |||
| BC132956.1 | - | 1 | 707 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00909| Gene Symbol: LINC00909 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024484.1 | 1 | 931 | |||
| GenBank mRNA | AK001853.1 | - | 1 | 678 | ||
| BC036588.2 | - | 1 | 922 | |||
| BC132930.1 | - | 1 | 707 | |||
| BC132956.1 | - | 1 | 707 | |||