siRNA Details
| Gene Symbol: SNAP47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017000231.1 | XP_016855720.1 | 3 | 2267 | ||
| XM_017000232.1 | XP_016855721.1 | 2 | 2073 | |||
| GenBank mRNA | AF370397.1 | - | 1 | 1373 | ||
| AK055266.1 | - | 3 | 2246 | |||
| AK093736.1 | - | 2 | 2076 | |||

SNAP47
Genomic Map