siRNA Details
| Gene Symbol: LOC441666 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024380.1 | 2 | 255 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA322429.1 | - | 2 | 184 | Chr. 10 - 42337880 - 42367974 [-] on Build GRCh38 | |

LOC441666
Genomic Map