siRNA Details
| Gene Symbol: FLJ26850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027257.1 | 3 | 1318 | |||
| GenBank mRNA | AK130360.1 | - | 3 | 1409 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ26850| Gene Symbol: FLJ26850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027257.1 | 3 | 1318 | |||
| GenBank mRNA | AK130360.1 | - | 3 | 1409 | ||