siRNA Details
| Gene Symbol: FLJ26850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027257.1 | 3 | 1280 | |||
| GenBank mRNA | AK130360.1 | - | 3 | 1371 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000527209.1 | - | 3 | 1371 | Chr. 19 - 50050589 - 50066793 [+] on Build GRCh38 | |

FLJ26850
Genomic Map