siRNA Details
| Gene Symbol: MIR1298 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_031578.1 | 1 | 15 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR1298| Gene Symbol: MIR1298 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_031578.1 | 1 | 15 | |||