siRNA Details
| Gene Symbol: LOC107987248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017025501.1 | XP_016880990.1 | 2 | 274 | ||
| Gene Symbol: TCAM1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002947.2 | 6 | 1734 | |||
| GenBank mRNA | AB026156.1 | - | 6 | 1734 | ||

LOC107987248
Genomic Map