siRNA Details
| Gene Symbol: TCAM1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002947.2 | 2 | 714 | |||
| GenBank mRNA | AB026156.1 | - | 2 | 714 | ||
| LC005096.1 | - | 4 | 805 | |||
| LC005097.1 | - | 4 | 574 | |||
| LC005098.1 | - | 3 | 542 | |||
| LC005099.1 | - | 4 | 659 | |||
| LC005100.1 | - | 4 | 590 | |||
| LC005101.1 | - | 4 | 556 | |||

TCAM1P
Genomic Map