siRNA Details
| Gene Symbol: FAM86B3P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024361.1 | 5 | 466 | |||
| NR_024362.1 | 5 | 563 | ||||
| NR_024363.1 | 4 | 461 | ||||
| GenBank mRNA | AK304567.1 | - | 5 | 535 | ||
| AK307256.1 | - | 4 | 431 | |||
| BC035792.1 | - | 5 | 441 | |||

FAM86B3P
Genomic Map