siRNA Details
| Gene Symbol: MAPT-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024560.1 | 1 | 316 | |||
| GenBank mRNA | AK091698.1 | - | 1 | 71 | ||
| DA504438.1 | - | 1 | 306 | |||
| DA789878.1 | - | 1 | 320 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAPT-IT1| Gene Symbol: MAPT-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024560.1 | 1 | 316 | |||
| GenBank mRNA | AK091698.1 | - | 1 | 71 | ||
| DA504438.1 | - | 1 | 306 | |||
| DA789878.1 | - | 1 | 320 | |||