siRNA Details
| Gene Symbol: LINC00310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027266.1 | 1 | 50 | |||
| NR_027267.1 | 1 | 50 | ||||
| GenBank mRNA | AF426258.1 | - | 1 | 50 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00310| Gene Symbol: LINC00310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027266.1 | 1 | 50 | |||
| NR_027267.1 | 1 | 50 | ||||
| GenBank mRNA | AF426258.1 | - | 1 | 50 | ||