siRNA Details
| Gene Symbol: ACBD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006717528.3 | XP_006717591.3 | 1 | 722 | ||
| XM_017016884.1 | XP_016872373.1 | 1 | 722 | |||
| XM_017016885.1 | XP_016872374.1 | 1 | 722 | |||
| XM_017016886.1 | XP_016872375.1 | 1 | 722 | |||
| XM_017016890.1 | XP_016872379.1 | 1 | 722 | |||
| XM_017016893.1 | XP_016872382.1 | 1 | 722 | |||
| XM_017016894.1 | XP_016872383.1 | 1 | 722 | |||
| Gene Symbol: LRRC37A6P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003525.2 | 1 | 4354 | |||
| GenBank mRNA | AK126872.1 | - | 1 | 2610 | ||

ACBD5
Genomic Map