siRNA Details
| Gene Symbol: LINC00313 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026863.1 | 3 | 394 | |||
| GenBank mRNA | AF426261.1 | - | 3 | 394 | ||
| BC107109.2 | - | 3 | 325 | |||
| Gene Symbol: LOC102724354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136540.1 | 4 | 355 | |||
| GenBank mRNA | HY162931.1 | - | 3 | 313 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX648824.1 | - | 1 | 64 | Chr. 21 - 43470433 - 43471592 [-] on Build GRCh38 | |

LINC00313
Genomic Map