siRNA Details
| Gene Symbol: MRPL42P5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002208.1 | 1 | 354 | |||
| Gene Symbol: C15orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289132.1 | NP_001276061.1 | 4 | 786 | ||
| GenBank mRNA | AK098781.1 | - | 4 | 721 | ||

MRPL42P5
Genomic Map