siRNA Details
| Gene Symbol: LINC01591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026706.2 | 6 | 1057 | |||
| NR_120395.1 | 6 | 1021 | ||||
| NR_120396.1 | 5 | 1040 | ||||
| GenBank mRNA | BC045817.1 | - | 6 | 1057 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01591| Gene Symbol: LINC01591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026706.2 | 6 | 1057 | |||
| NR_120395.1 | 6 | 1021 | ||||
| NR_120396.1 | 5 | 1040 | ||||
| GenBank mRNA | BC045817.1 | - | 6 | 1057 | ||