siRNA Details
| Gene Symbol: LINC01591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026706.2 | 4 | 637 | |||
| NR_120395.1 | 4 | 601 | ||||
| NR_120396.1 | 3 | 620 | ||||
| GenBank mRNA | BC045817.1 | - | 4 | 637 | ||
| BC117350.1 | - | 4 | 530 | |||
| BC117352.1 | - | 4 | 531 | |||
| BC144006.1 | - | 4 | 495 | |||
| BC171736.1 | - | 3 | 514 | |||

LINC01591
Genomic Map