siRNA Details
Gene Symbol: ZNF252P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023392.1 | 5 | 2954 | |||
GenBank mRNA | AK027662.1 | - | 1 | 988 | ||
AK128723.1 | - | 1 | 1278 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF252P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023392.1 | 5 | 2954 | |||
GenBank mRNA | AK027662.1 | - | 1 | 988 | ||
AK128723.1 | - | 1 | 1278 |