siRNA Details
| Gene Symbol: ZNF252P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_023392.1 | 5 | 3341 | |||
| GenBank mRNA | AK027662.1 | - | 1 | 1368 | ||
| AK128723.1 | - | 1 | 1666 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF252P| Gene Symbol: ZNF252P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_023392.1 | 5 | 3341 | |||
| GenBank mRNA | AK027662.1 | - | 1 | 1368 | ||
| AK128723.1 | - | 1 | 1666 | |||