siRNA Details
| Gene Symbol: TMED10P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002807.4 | 1 | 3508 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | HY307714.1 | - | 1 | 421 | Chr. 8 - 144997968 - 144998455 [+] on Build GRCh38 | |

TMED10P1
Genomic Map