siRNA Details
| Gene Symbol: ZNF252P-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026974.1 | 1 | 2760 | |||
| GenBank mRNA | AK092777.1 | - | 1 | 2585 | ||
| BC046439.1 | - | 1 | 2760 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ185413.1 | - | 1 | 367 | Chr. 8 - 145005204 - 145005938 [+] on Build GRCh38 | |

ZNF252P-AS1
Genomic Map