siRNA Details
| Gene Symbol: ZNF252P-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026974.1 | 1 | 2982 | |||
| GenBank mRNA | BX104600.1 | - | 1 | 209 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF252P-AS1| Gene Symbol: ZNF252P-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026974.1 | 1 | 2982 | |||
| GenBank mRNA | BX104600.1 | - | 1 | 209 | ||