siRNA Details
| Gene Symbol: PDIA3P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002305.1 | 1 | 1542 | |||
| GenBank mRNA | AK295985.1 | - | 1 | 1427 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PDIA3P1| Gene Symbol: PDIA3P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002305.1 | 1 | 1542 | |||
| GenBank mRNA | AK295985.1 | - | 1 | 1427 | ||