siRNA Details
| Gene Symbol: MIR1236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_031601.1 | 1 | 49 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR1236| Gene Symbol: MIR1236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_031601.1 | 1 | 49 | |||