siRNA Details
| Gene Symbol: HSP90B3P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY956769.1 | - | 3 | 713 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HSP90B3P| Gene Symbol: HSP90B3P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY956769.1 | - | 3 | 713 | ||