siRNA Details
| Gene Symbol: FAM183CP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024473.1 | 1 | 834 | |||
| GenBank mRNA | AL833525.1 | - | 1 | 528 | ||
| BC028745.1 | - | 1 | 838 | |||
| BC045833.1 | - | 1 | 833 | |||
| BC126344.1 | - | 1 | 834 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM183CP| Gene Symbol: FAM183CP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024473.1 | 1 | 834 | |||
| GenBank mRNA | AL833525.1 | - | 1 | 528 | ||
| BC028745.1 | - | 1 | 838 | |||
| BC045833.1 | - | 1 | 833 | |||
| BC126344.1 | - | 1 | 834 | |||