siRNA Details
| Gene Symbol: LOC440910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030728.1 | 1 | 28 | |||
| GenBank mRNA | AK091798.1 | - | 1 | 28 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA584196.1 | - | 1 | 28 | Chr. 2 - 131279290 - 131296893 [+] on Build GRCh38 | |

LOC440910
Genomic Map