siRNA Details
| Gene Symbol: LOC440910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030728.1 | 4 | 545 | |||
| GenBank mRNA | AK091798.1 | - | 4 | 545 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC440910| Gene Symbol: LOC440910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030728.1 | 4 | 545 | |||
| GenBank mRNA | AK091798.1 | - | 4 | 545 | ||